Assay Details
| Gene Symbol: Gm38483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK053109.1 | - | 976 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NFKB inhibitor interacting Ras-like protein 2 | Nkiras2 |
2410003M04Rik, 4930527H08Rik, D630018G21Rik, KBRAS2 |
-63.2kb | Sense | Chr. 11 - 100622955 - 100627602 on NCBI Build GRCm38 |
| Downstream | DEXH (Asp-Glu-X-His) box polypeptide 58 | Dhx58 |
B430001I08Rik, D11Lgp2e, LPG2, Lgp2, RLR-3 |
3.8kb | Anti-sense | Chr. 11 - 100694884 - 100704271 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm38483
Genomic Map