Assay Details
| Gene Symbol: Gm41959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076660.1 | - | 277 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 4258 | Gm4258 | Anti-sense | Chr. 1 - 137898476 - 137960785 on NCBI Build GRCm38 | ||
| Downstream | microRNA 181a-1 | Mir181a-1 |
Mirn181a-1, Mirn213, miR-213, mir-181a-1, mmu-mir-213 |
35.6kb | Anti-sense | Chr. 1 - 137966455 - 137966541 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm41959
Genomic Map