Assay Details
Gene Symbol: Gm13752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040370.1 | 2 - 3 | 477 | 195 | ||
GenBank mRNA | AK139560.1 | - | 2 - 3 | 438 | 195 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
DnaJ (Hsp40) homolog, subfamily C, member 10 |
Dnajc10 |
1200006L06Rik, D2Ertd706e, ERdj5, JPDI |
-40.8kb | Sense | Chr. 2 - 80315466 - 80354055 on NCBI Build GRCm38 |
Downstream | frizzled-related protein | Frzb |
Frp, Sfrp3, frezzled, fritz, frzb-1 |
16.7kb | Anti-sense | Chr. 2 - 80411970 - 80447396 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205441 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 266015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374517 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374515 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374473 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374533 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374498 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02195119 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374431 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374493 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374537 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02209289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374419 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374512 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374531 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01484852 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02174882 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01916195 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374492 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02144424 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02144427 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374414 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374475 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374485 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374427 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374534 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374417 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01916189 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374476 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374543 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374453 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374522 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374501 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02195107 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374442 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374436 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374474 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374438 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374444 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374505 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374448 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374502 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01226302 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374524 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374446 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374486 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374507 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374510 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374504 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01884986 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374447 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374451 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374525 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374440 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374469 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02195112 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374489 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374433 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374415 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01445653 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374508 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374496 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02174888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374499 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374472 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374483 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374541 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374490 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01916196 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374467 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21374460 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0200009430.mm.1 | NR_040370.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009430.mm.1 | NR_040370.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009430.mm.1 | TC0200001172.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200009432.mm.1 | NR_040370.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009432.mm.1 | NR_040370.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009432.mm.1 | TC0200001172.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200009431.mm.1 | NR_040370.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009431.mm.1 | NR_040370.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009431.mm.1 | TC0200001172.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200009433.mm.1 | NR_040370.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009433.mm.1 | NR_040370.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009433.mm.1 | TC0200001172.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200009429.mm.1 | NR_040370.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009429.mm.1 | NR_040370.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009429.mm.1 | TC0200001172.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 726785 | - | - |
