Assay Details
Gene Symbol: Gm33180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK139643.1 | - | 2 - 3 | 360 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ariadne ubiquitin-conjugating enzyme E2 binding protein homolog 1 (Drosophila) | Arih1 |
AU021774, Ari, Ari1, Hari, Hhari, Ubch7bp, Uip77 |
-12.7kb | Sense | Chr. 9 - 59388554 - 59486660 on NCBI Build GRCm38 |
Downstream | transmembrane protein 202 | Tmem202 | 4930425N13Rik | 7.5kb | Sense | Chr. 9 - 59518685 - 59525501 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
