Assay Details
Gene Symbol: Gm16170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK139983.1 | - | 1 - 2 | 760 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tripartite motif-containing 41 | Trim41 |
AW552703, BC020156, R75223, RINCK |
-29.9kb | Sense | Chr. 11 - 48806403 - 48817765 on NCBI Build GRCm38 |
Downstream | immunity-related GTPase family M member 1 | Irgm1 |
Ifggd3, Ifi1, Iigp3, Iipg3, Irgm, LRG-47 |
11.8kb | Sense | Chr. 11 - 48865245 - 48871415 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
