Assay Details
Gene Symbol: Gm15083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK140501.1 | - | 2 - 3 | 208 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
ATPase, Ca++ transporting, plasma membrane 2 |
Atp2b2 |
D6Abb2e, Gena300, PMCA2, Tmy, dfw, jog, wms, wri |
-7.7kb | Anti-sense | Chr. 6 - 113743842 - 114042613 on NCBI Build GRCm38 |
Downstream |
solute carrier family 6 (neurotransmitter transporter, GABA), member 11 |
Slc6a11 |
D930045G19Rik, E130202I16Rik, GAT4, Gabt4, Gat3 |
79.0kb | Sense | Chr. 6 - 114131241 - 114249886 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
