Assay Details
| Gene Symbol: LOC105246847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK149321.1 | - | 1 - 2 | 288 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 192 | Mir192 |
Mirn192, mir-192, mmu-mir-192 |
-3.9kb | Anti-sense | Chr. 19 - 6264844 - 6264932 on NCBI Build GRCm38 |
| Downstream | EH-domain containing 1 | Ehd1 |
AA409636, Past1, RME-1 |
318.0 | Anti-sense | Chr. 19 - 6276896 - 6300096 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105246847
Genomic Map