Assay Details
| Gene Symbol: 1500017E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK149390.1 | - | 2 - 3 | 91 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HECT domain containing 2 | Hectd2 |
4921524L07, A630025O09Rik, AW212605 |
-4.1kb | Anti-sense | Chr. 19 - 36553969 - 36621135 on NCBI Build GRCm38 |
| Downstream |
protein phosphatase 1, regulatory (inhibitor) subunit 3C |
Ppp1r3c |
PTG, Ppp1r5 |
105.8kb | Sense | Chr. 19 - 36731731 - 36736604 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1500017E21Rik
Genomic Map