Assay Details
| Gene Symbol: Gm16892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK149401.1 | - | 2 - 3 | 177 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
nitrilase family, member 2 |
Nit2 |
1190017B19Rik, D16Ertd502e |
-1.9kb | Anti-sense | Chr. 16 - 57156665 - 57167332 on NCBI Build GRCm38 |
| Downstream | transmembrane protein 30C | Tmem30c |
4933401B01Rik, 4933409A18Rik, Cdc50c |
94.6kb | Anti-sense | Chr. 16 - 57266138 - 57301893 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16892
Genomic Map