Assay Details
| Gene Symbol: Gm36536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK158658.1 | - | 2 - 3 | 226 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phospholipase D family, member 5 |
Pld5 | B230365F16Rik | -174.0kb | Sense | Chr. 1 - 175962301 - 176280017 on NCBI Build GRCm38 |
| Downstream | centrosomal protein 170 | Cep170 |
4933426L22Rik, A330004A13Rik, AI195353 |
215.4kb | Sense | Chr. 1 - 176733653 - 176814173 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm36536
Genomic Map