Assay Details
| Gene Symbol: Gm33027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC092221.1 | - | 98 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphatidic acid phosphatase type 2 domain containing 1A | Ppapdc1a |
C030048B12Rik, Gm1090 |
-10.7kb | Anti-sense | Chr. 7 - 129257094 - 129391307 on NCBI Build GRCm38 |
| Downstream | WD repeat domain 11 | Wdr11 |
2900055P10Rik, AW489876, Brwd2, mKIAA1351 |
188.6kb | Anti-sense | Chr. 7 - 129591714 - 129635738 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm33027
Genomic Map