Assay Details
Gene Symbol: Gm29999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC100375.1 | - | 1 - 2 | 92 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | neuropeptide Y receptor Y2 | Npy2r | NPY2-R | -111.5kb | Sense | Chr. 3 - 82538383 - 82548989 on NCBI Build GRCm38 |
Downstream | RNA binding motif protein 46 | Rbm46 |
EG633285, ENSMUSG00000033882, Gm761 |
171.6kb | Sense | Chr. 3 - 82836521 - 82876533 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
