Assay Details
| Gene Symbol: Gm2137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082723.1 | - | 1 | 1357 | 76 | |
| BB396393.2 | - | 1 | 502 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 5108 | Mir5108 |
mmu-mir-5108, mu-mir-5108 |
-8.5kb | Sense | Chr. 10 - 61774737 - 61774821 on NCBI Build GRCm38 |
| Downstream |
collagen, type XIII, alpha 1 |
Col13a1 | 53.4kb | Anti-sense | Chr. 10 - 61838235 - 61979209 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm2137
Genomic Map