Assay Details
| Gene Symbol: Gm8273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK084737.1 | - | 1 | 351 | 99 | |
| BB479211.2 | - | 1 | 362 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB2A, member RAS oncogene family |
Rab2a |
9330148M11Rik, C80220, Rab2 |
-39.3kb | Anti-sense | Chr. 4 - 8535586 - 8607702 on NCBI Build GRCm38 |
| Downstream | chromodomain helicase DNA binding protein 7 | Chd7 |
A730019I05Rik, Cycn, Cyn, Dz, Edy, Flo, GENA 47, GENA 60, Gena 52, Lda, Mt, Obt, Todo, WBE1, Whi, metis |
43.0kb | Anti-sense | Chr. 4 - 8690406 - 8868449 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm8273
Genomic Map