Assay Details
| Gene Symbol: Gm15759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028685.1 | - | 563 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | aldehyde oxidase 2 | Aox2 |
Aoh3, Aox3l1 |
Anti-sense | Chr. 1 - 58276141 - 58379264 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 39644 |
Gm39644 | 67.5kb | Sense | Chr. 1 - 58366418 - 58392543 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm15759
Genomic Map