Assay Details
| Gene Symbol: Gm13599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029730.1 | - | 3 - 4 | 3700 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 1322 | Gm1322 | -123.4kb | Anti-sense | Chr. 2 - 67173834 - 67186212 on NCBI Build GRCm38 | |
| Downstream | xin actin-binding repeat containing 2 | Xirp2 |
2310003D02Rik, 2310008C07Rik, A530024P18Rik, AI452089, Cmya3, Gm352, Xin2 |
104.1kb | Anti-sense | Chr. 2 - 67446000 - 67526620 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm13599
Genomic Map