Assay Details
Gene Symbol: Gm29886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK131767.1 | - | 1 - 2 | 697 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | adhesion regulating molecule 1 | Adrm1 |
1110063P18Rik, 2510006J17Rik, AA408205, ARM-1, AU043535, Arm1, Gp110, Rpn13 |
Anti-sense | Chr. 2 - 180171490 - 180176286 on NCBI Build GRCm38 | |
Downstream |
laminin, alpha 5 |
Lama5 |
AA408760, AA408762, AI853660, [a]5, laminin-511, mKIAA0533 |
4.5kb | Sense | Chr. 2 - 180176373 - 180225905 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
