Assay Details
| Gene Symbol: Gm36615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK135696.1 | - | 2 - 3 | 100 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane and immunoglobulin domain containing 1 | Tmigd1 |
2010002A20Rik, Tmigd |
-2.0kb | Anti-sense | Chr. 11 - 76891246 - 76916587 on NCBI Build GRCm38 |
| Downstream | bleomycin hydrolase | Blmh |
AI035728, Bh, Bmh |
25.1kb | Anti-sense | Chr. 11 - 76945656 - 76987389 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm36615
Genomic Map