Assay Details
Gene Symbol: Gm30052 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK135717.1 | - | 2 - 3 | 181 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | synaptonemal complex central element protein 1 like | Syce1l |
4930481F22Rik, MMRP, PSESS, mmrp2 |
-12.4kb | Anti-sense | Chr. 8 - 113643213 - 113655533 on NCBI Build GRCm38 |
Downstream |
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 18 |
Adamts18 |
9630038L21, ADAMTS21, E130314N14Rik |
22.8kb | Sense | Chr. 8 - 113697123 - 113855050 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
