Assay Details
| Gene Symbol: Gm33354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK135929.1 | - | 2 - 3 | 376 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pogo transposable element with KRAB domain | Pogk |
9130401E23Rik, BASS2, mKIAA1513 |
-38.4kb | Sense | Chr. 1 - 166393610 - 166409858 on NCBI Build GRCm38 |
| Downstream | predicted gene 4846 | Gm4846 |
EG226601, Fmo13 |
29.1kb | Sense | Chr. 1 - 166482599 - 166506135 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm33354
Genomic Map