Assay Details
Gene Symbol: Gm15614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK136552.1 | - | 2 - 3 | 375 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
protein phosphatase 1, catalytic subunit, beta isoform |
Ppp1cb | 1200010B19 | -15.6kb | Anti-sense | Chr. 5 - 32458848 - 32493712 on NCBI Build GRCm38 |
Downstream | Yamaguchi sarcoma viral (v-yes) oncogene homolog 1 | Yes1 |
Yes, p61-Yes |
29.0kb | Anti-sense | Chr. 5 - 32611171 - 32687066 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
