Assay Details
Gene Symbol: Gm33994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK137777.1 | - | 1 - 2 | 2188 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | kinesin family member 14 | Kif14 |
D1Ertd367e, E130203M01 |
-21.0kb | Anti-sense | Chr. 1 - 136466290 - 136555938 on NCBI Build GRCm38 |
Downstream | zinc finger protein 281 | Zfp281 | Znf281 | 45.4kb | Anti-sense | Chr. 1 - 136624901 - 136630391 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
