Assay Details
| Gene Symbol: AI463170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046044.1 | 1 - 2 | 222 | 91 | ||
| GenBank mRNA | AA042017.1 | - | 1 - 2 | 222 | 91 | |
| AK144388.1 | - | 1 - 2 | 113 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 5135 | Mir5135 |
mir-5135, mmu-mir-5135, mu-mir-5135 |
-14.2kb | Sense | Chr. 12 - 76533134 - 76533212 on NCBI Build GRCm38 |
| Downstream |
spectrin beta, erythrocytic |
Sptb |
AI842465, D330027P03Rik, Gm1301, Spnb-1, Spnb1, ja, jaundiced, mKIAA4219 |
32.7kb | Sense | Chr. 12 - 76580488 - 76710547 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

AI463170
Genomic Map