Assay Details
Gene Symbol: Gm36229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK144827.1 | - | 2 - 3 | 155 | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | centrosomal protein 85-like | Cep85l |
ENSMUSG00000038594, Gm9766 |
-40.8kb | Anti-sense | Chr. 10 - 53276799 - 53380701 on NCBI Build GRCm38 |
Downstream |
predicted gene, 20597 |
Gm20597 | B230314j19 | 74.2kb | Anti-sense | Chr. 10 - 53498443 - 53518989 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
