Assay Details
| Gene Symbol: Gm41790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155068.1 | - | 3 - 4 | 280 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glucosamine-6-phosphate deaminase 1 pseudogene | Gm2176 | A430076E10Rik | -27.3kb | Sense | Chr. 18 - 80541892 - 80544546 on NCBI Build GRCm38 |
| Downstream |
nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 1 |
Nfatc1 |
2210017P03Rik, AI449492, AV076380, NF-ATc, NFAT2, NFATc, Nfatcb |
32.3kb | Sense | Chr. 18 - 80606205 - 80713071 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm41790
Genomic Map