Assay Details
| Gene Symbol: Gm19863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK157548.1 | - | 1 | 1952 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
mannoside acetylglucosaminyltransferase 4, isoenzyme A |
Mgat4a |
9530018I07Rik, GnT-IVa, glcNAc-T-IVa |
-153.4kb | Sense | Chr. 1 - 37439340 - 37541047 on NCBI Build GRCm38 |
| Downstream | testis specific 10 | Tsga10 |
4933432N21Rik, AI843383, Gm217, Mtsga10 |
31.5kb | Sense | Chr. 1 - 37728144 - 37866739 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm19863
Genomic Map