Assay Details
Gene Symbol: Gm3740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK158189.1 | - | 1 - 2 | 132 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 31359 |
Gm31359 | -153.9kb | Sense | Chr. 3 - 85391545 - 85402099 on NCBI Build GRCm38 | |
Downstream |
glutamyl-tRNA(Gln) amidotransferase, subunit B |
Gatb |
9430026F02Rik, Pet112, Pet112l |
157.0 | Anti-sense | Chr. 3 - 85574102 - 85655660 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
