Assay Details
Gene Symbol: Gm16630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK158926.1 | - | 4 - 5 | 1026 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 746 | Zfp746 |
2810407L07Rik, AI317225, Znf746 |
-43.9kb | Sense | Chr. 6 - 48062381 - 48086990 on NCBI Build GRCm38 |
Downstream | KRAB-A domain containing 1 | Krba1 |
A930040G15Rik, AI448780 |
253.2kb | Anti-sense | Chr. 6 - 48395586 - 48419855 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
