Assay Details
| Gene Symbol: LOC105244195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079188.1 | - | 1 - 2 | 195 | 66 | |
| AK162487.1 | - | 1 - 2 | 192 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rap guanine nucleotide exchange factor (GEF) 4 | Rapgef4 |
1300003D15Rik, 5730402K07Rik, 6330581N18Rik, EPAC 2, Epac-2, Epac2, mKIAA4040 |
Anti-sense | Chr. 2 - 71981215 - 72257474 on NCBI Build GRCm38 | |
| Downstream | sterile alpha motif and leucine zipper containing kinase AZK | Zak |
AV006891, B230120H23Rik, MLTK, MLTKalpha, MLTKbeta |
53.3kb | Anti-sense | Chr. 2 - 72285637 - 72442610 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

LOC105244195
Genomic Map