Assay Details
| Gene Symbol: Gm15622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016283.1 | - | 2 - 3 | 172 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | junction-mediating and regulatory protein | Jmy | -125.6kb | Anti-sense | Chr. 13 - 93430097 - 93499808 on NCBI Build GRCm38 | |
| Downstream | betaine-homocysteine methyltransferase 2 | Bhmt2 |
C81077, D13Ucla2 |
27.5kb | Anti-sense | Chr. 13 - 93656097 - 93674302 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512447 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947793 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99662 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147765 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922515 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131610 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687874 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71011 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156701 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07955818 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07955797 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07955831 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07955797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10782572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10486989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11056413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11056398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10624047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10921640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10872958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10514806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11056411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10624054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10786137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10876962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11011546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11056402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11056397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10486993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29229986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230028 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1431342_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431342_at | - | - |
| Mouse Genome 430 2.0 Array | 1431342_PM_at | - | - |
| Mouse Expression Set 430 | 1431342_PM_at | - | - |
| Mouse Expression Set 430 | 1431342_at | - | - |

Gm15622
Genomic Map