Assay Details
| Gene Symbol: Gm26839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131831.1 | - | 1 - 2 | 121 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | forkhead box N3 | Foxn3 |
5430426H20Rik, AA589593, AW556347, Ches1, Ches1l, HTLFL1 |
-216.0 | Anti-sense | Chr. 12 - 99190081 - 99563625 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700064M15 gene | 1700064M15Rik | 60.8kb | Anti-sense | Chr. 12 - 99626053 - 99627974 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm26839
Genomic Map