Assay Details
Gene Symbol: Gm36173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK136704.1 | - | 1 - 2 | 323 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
solute carrier family 35, member F1 |
Slc35f1 |
Duf6, E130016L02 |
-123.6kb | Sense | Chr. 10 - 52690501 - 53111622 on NCBI Build GRCm38 |
Downstream | centrosomal protein 85-like | Cep85l |
ENSMUSG00000038594, Gm9766 |
22.9kb | Anti-sense | Chr. 10 - 53276799 - 53380701 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
