Assay Details
| Gene Symbol: Gm34006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK145144.1 | - | 1 - 2 | 343 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sex comb on midleg-like 4 (Drosophila) | Scml4 |
9330161D17, mFLJ00197 |
Anti-sense | Chr. 10 - 42860317 - 42960782 on NCBI Build GRCm38 | |
| Downstream | predicted gene 5079 | Gm5079 | C230040G06 | 106.6kb | Sense | Chr. 10 - 42971283 - 42992164 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm34006
Genomic Map