Assay Details
Gene Symbol: Gm26802 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK132834.1 | - | 1 | 1352 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | reticulon 2 (Z-band associated protein) | Rtn2 |
MMS10-P, Ms10p, NSPLI, Nspl1 |
-2.9kb | Anti-sense | Chr. 7 - 19282624 - 19296164 on NCBI Build GRCm38 |
Downstream | nuclear encoded tRNA selenocysteine 2 (anticodon TCA) | n-Ts2 |
Trnau1, Trsp, tRNA-Sec |
474.0 | Anti-sense | Chr. 7 - 19301244 - 19301329 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
