Assay Details
| Gene Symbol: 2810417H13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK134432.1 | - | 2 | 252 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thyroid hormone receptor interactor 4 | Trip4 |
4930558E03Rik, ASC-1, Asc1, BB191711 |
Anti-sense | Chr. 9 - 65828926 - 65908809 on NCBI Build GRCm38 | |
| Downstream |
casein kinase 1, gamma 1 |
Csnk1g1 | 9130020E21Rik | 15.8kb | Sense | Chr. 9 - 65908962 - 66045015 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2810417H13Rik
Genomic Map