Assay Details
Gene Symbol: Gm34816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK136242.1 | - | 3 | 357 | 113 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4933436E23 gene | 4933436E23Rik | -10.8kb | Sense | Chr. 1 - 139379452 - 139402852 on NCBI Build GRCm38 | |
Downstream | zinc finger and BTB domain containing 41 homolog | Zbtb41 |
8430415N23Rik, 9430031N01, 9830132G07Rik, AI316857 |
7.9kb | Sense | Chr. 1 - 139421719 - 139453007 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
