Assay Details
Gene Symbol: LOC102639173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK138181.1 | - | 2 | 1078 | 104 | |
AK148373.1 | - | 2 | 450 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | inositol polyphosphate phosphatase-like 1 | Inppl1 |
51C, SHIP2 |
-576.0 | Anti-sense | Chr. 7 - 101822632 - 101838226 on NCBI Build GRCm38 |
Downstream | folate receptor 2 (fetal) | Folr2 |
FBP2, FR-P3, FR-beta, Folbp-2, Folbp2 |
969.0 | Anti-sense | Chr. 7 - 101839988 - 101857190 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
