Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200006803.mm.1 |
NM_001008506.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006803.mm.1 |
NM_001008506.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006803.mm.1 |
NM_001160107.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006803.mm.1 |
NM_001160107.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006803.mm.1 |
NM_001160108.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006803.mm.1 |
NM_001160108.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006803.mm.1 |
NM_029334.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006803.mm.1 |
NM_029334.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006803.mm.1 |
NR_027648.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006803.mm.1 |
NR_027648.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006803.mm.1 |
TC1200000970.mm.1 |
MTA TC |