Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10478916 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10478915 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10478914 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10478921 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10478919 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10478917 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10478920 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10478918 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1439242_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1428924_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10478913 |
- |
- |
Mouse Genome 430 2.0 Array |
1439242_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1439242_at |
- |
- |
Mouse Genome 430 2.0 Array |
1428924_at |
- |
- |
Mouse Genome 430 2.0 Array |
1428924_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
69372_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
69372_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0200021460.mm.1 |
NM_001160330.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021460.mm.1 |
NM_001160330.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021460.mm.1 |
TC0200002608.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1439242_PM_at |
- |
- |
Mouse Expression Set 430 |
1428924_at |
- |
- |
Mouse Expression Set 430 |
1428924_PM_at |
- |
- |
Mouse Expression Set 430 |
1439242_at |
- |
- |
Murine Genome U74v2 Set |
139229_r_at |
- |
- |
Murine Genome U74v2 Set |
139229_PM_r_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1439242_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1428924_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1428924_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1439242_PM_at |
- |
- |