Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400004512.mm.1 |
NM_001162863.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004512.mm.1 |
NM_001162863.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004512.mm.1 |
TC1400000576.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004515.mm.1 |
NM_001162863.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004515.mm.1 |
NM_001162863.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004515.mm.1 |
NM_029145.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004515.mm.1 |
NM_029145.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004515.mm.1 |
TC1400000576.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004516.mm.1 |
NM_001162863.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004516.mm.1 |
NM_001162863.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004516.mm.1 |
NM_029145.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004516.mm.1 |
NM_029145.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004516.mm.1 |
TC1400000576.mm.1 |
MTA TC |