Assay Details
| Gene Symbol: Tmem170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163572.1 | NP_001157044.1 | 2 - 3 | 271 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978336 | NM_001163572.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 260139 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833692 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641869 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318408 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30378 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218875 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597408 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221551 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 296112 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628048 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192919 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 214541 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561678 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944667 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36733 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 507636 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858790 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597631 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19616 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 142862 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 15126 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978336 | NM_001163572.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10856883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10507720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11078572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10856891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11078593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11078582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10856889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28979230 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1434556_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404763 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300002825.mm.1 | NM_001163572.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002825.mm.1 | NM_001163572.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002825.mm.1 | TC1300000469.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002826.mm.1 | NM_001163572.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002826.mm.1 | NM_001163572.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002826.mm.1 | TC1300000469.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002827.mm.1 | NM_001163572.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002827.mm.1 | NM_001163572.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002827.mm.1 | TC1300000469.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002828.mm.1 | NM_001163572.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002828.mm.1 | NM_001163572.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002828.mm.1 | TC1300000469.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434556_PM_at | - | - |
| Mouse Expression Set 430 | 1434556_at | - | - |

Tmem170b
Genomic Map