Assay Details
| Gene Symbol: Apol6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163621.1 | NP_001157093.1 | 1 - 2 | 99 | 68 | |
| GenBank mRNA | AK144563.1 | - | 1 - 2 | 100 | 68 | |
| BC113126.1 | - | 1 - 2 | 445 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001163621.1 transcripts.
Set Membership: |
|

Apol6
Genomic Map