Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400016704.mm.1 |
NM_001164289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016704.mm.1 |
NM_001164289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016704.mm.1 |
TC1400002870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016703.mm.1 |
NM_001164289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016703.mm.1 |
NM_001164289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016703.mm.1 |
TC1400002870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016705.mm.1 |
NM_001164289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016705.mm.1 |
NM_001164289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016705.mm.1 |
TC1400002870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016706.mm.1 |
NM_001164289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016706.mm.1 |
NM_001164289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016706.mm.1 |
TC1400002870.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016707.mm.1 |
NM_001164289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016707.mm.1 |
NM_001164289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016707.mm.1 |
TC1400002870.mm.1 |
MTA TC |