Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10600587 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1431445_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10600586 |
- |
- |
Mouse Genome 430 2.0 Array |
1431445_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1431445_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
70864_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
70864_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0X00005322.mm.1 |
NM_001164739.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005322.mm.1 |
NM_001164739.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005322.mm.1 |
TC0X00000791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005323.mm.1 |
NM_001164739.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005323.mm.1 |
NM_001164739.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005323.mm.1 |
TC0X00000791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005324.mm.1 |
NM_001164739.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005324.mm.1 |
NM_001164739.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005324.mm.1 |
TC0X00000791.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1431445_at |
- |
- |
Mouse Expression Set 430 |
1431445_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1431445_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1431445_PM_at |
- |
- |