Assay Details
Gene Symbol: Bend4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164806.1 | NP_001158278.1 | 1 - 2 | 732 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2199118 | AK052318.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2199118 | NM_001164806.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148844 | NM_001164806.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 474928 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231047 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478047 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 20962 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148844 | NM_001164806.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2199118 | AK052318.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2199118 | NM_001164806.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05602119 | - | - |
Whole Mouse Genome G4122A | A_52_P505038 | AK052318.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P505038 | BB449568.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P505038 | NM_001164806.1 | mouse_transcriptome |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767612 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767573 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767636 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04797194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767590 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767587 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767596 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04446659 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04867920 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04428457 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04446661 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767638 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767647 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04867928 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04207929 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767581 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767652 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767616 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767592 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767656 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04207936 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767617 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767630 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767631 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767611 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04446660 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767618 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04797208 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04207933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767624 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04146509 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767613 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767651 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767575 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04830576 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767585 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767661 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767645 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767574 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04207945 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767657 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767593 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767632 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04702106 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04428450 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23767629 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1439523_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456795_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10530310 | - | - |
Mouse Genome 430 2.0 Array | 1439523_at | - | - |
Mouse Genome 430 2.0 Array | 1456795_at | - | - |
Mouse Genome 430 2.0 Array | 1439523_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456795_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500022474.mm.1 | TC0500002526.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500022474.mm.1 | NM_001164806.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022474.mm.1 | NM_001164806.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022475.mm.1 | TC0500002526.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500022475.mm.1 | NM_001164806.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022475.mm.1 | NM_001164806.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022478.mm.1 | TC0500002526.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500022478.mm.1 | NM_001164806.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022478.mm.1 | NM_001164806.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022473.mm.1 | TC0500002526.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500022473.mm.1 | NM_001164806.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022473.mm.1 | NM_001164806.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022476.mm.1 | TC0500002526.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500022476.mm.1 | NM_001164806.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022476.mm.1 | NM_001164806.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022477.mm.1 | TC0500002526.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500022477.mm.1 | NM_001164806.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500022477.mm.1 | NM_001164806.1 | Refseq |
Mouse Expression Set 430 | 1439523_at | - | - |
Mouse Expression Set 430 | 1456795_at | - | - |
Mouse Expression Set 430 | 1456795_PM_at | - | - |
Mouse Expression Set 430 | 1439523_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1439523_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456795_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456795_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1439523_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 879301 | - | - |
