Assay Details
| Gene Symbol: Fam89b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023166.2 | NP_075655.2 | 1 - 2 | 428 | 73 | |
| GenBank mRNA | AF212921.1 | - | 1 - 2 | 395 | 73 | |
| AK003883.1 | - | 1 - 2 | 417 | 73 | ||
| BC056166.1 | - | 1 - 2 | 413 | 5715229 | 73 | |
| CJ132526.1 | - | 1 - 2 | 73 | |||
More Information
Set Membership: |
|

Fam89b
Genomic Map