Assay Details
Gene Symbol: Fam160b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194345.1 | NP_919326.1 | 1 - 2 | 93 | 81 | |
GenBank mRNA | BC050861.1 | - | 1 - 2 | 94 | 81 | |
BU152183.1 | - | 1 - 2 | 93 | 81 | ||
CF170130.1 | - | 1 - 2 | 49 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
