Assay Details
Gene Symbol: Gm14446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101605.1 | NP_001095075.1 | 2 | 2147 | 110 | |
NM_001110517.1 | NP_001103987.1 | 2 | 2228 | 110 | ||
GenBank mRNA | AK090072.1 | - | 3 | 2168 | 110 | |
BQ553020.1 | - | 1 | 110 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500082 | BX515248.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500082 | NM_001110517.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500082 | AK090072.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500082 | BQ553020.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500082 | NM_001101605.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500082 | BX515248.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500082 | NM_001110517.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500082 | AK090072.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500082 | BQ553020.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P500082 | NM_001101605.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552374 | AK087331.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552374 | BC048932.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 256278 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 845561 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288039 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111597 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 745025 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267207 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 615570 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 363032 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821507 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499588 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 942804 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925414 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 540266 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164078 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413583 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379371 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77593 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163712 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31555 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156686 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500082 | BX515248.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500082 | NM_001110517.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500082 | AK090072.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500082 | BQ553020.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500082 | NM_001101605.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500082 | BX515248.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500082 | NM_001110517.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500082 | AK090072.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500082 | BQ553020.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P500082 | NM_001101605.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09158501 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09158632 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09158649 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09158532 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09158513 | - | - |
Whole Mouse Genome G4122A | A_51_P500082 | BX515248.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P500082 | NM_001110517.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P500082 | AK090072.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P500082 | BQ553020.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P500082 | NM_001101605.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P500082 | BX515248.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P500082 | NM_001110517.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P500082 | AK090072.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P500082 | BQ553020.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P500082 | NM_001101605.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P294064 | NM_001110517.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P294064 | AA175618.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P294064 | NM_001101605.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140944 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09158501 | - | - |
Agilent Mouse cDNA Microarray (G4104A) | 619141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32081127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081110 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081107 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081102 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081104 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081128 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14212995 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14440391 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081124 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081111 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14333548 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081098 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14456593 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081106 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32081101 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14145839 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1435529_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1435529_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1435529_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 667373_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 667373_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1900012437.mm.1 | NM_001101605.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900012437.mm.1 | NM_001101605.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900012437.mm.1 | TC1900001422.mm.1 | MTA TC |
Mouse Expression Set 430 | 1435529_PM_at | - | - |
Mouse Expression Set 430 | 1435529_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435529_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435529_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 643417 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 103610008 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 103610008 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103610008 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103610008 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1229197 | - | - |
