Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00003868.mm.1 |
NM_001166619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003868.mm.1 |
NM_001166619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003868.mm.1 |
NM_001166620.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003868.mm.1 |
NM_001166620.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003868.mm.1 |
NM_174993.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003868.mm.1 |
NM_174993.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003868.mm.1 |
TC0X00000614.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003870.mm.1 |
NM_001166619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003870.mm.1 |
NM_001166619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003870.mm.1 |
NM_174993.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003870.mm.1 |
NM_174993.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003870.mm.1 |
TC0X00000614.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003871.mm.1 |
NM_174993.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003871.mm.1 |
NM_174993.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003871.mm.1 |
TC0X00000614.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003869.mm.1 |
NM_001166619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003869.mm.1 |
NM_001166619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003869.mm.1 |
NM_001166620.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003869.mm.1 |
NM_001166620.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003869.mm.1 |
NM_174993.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003869.mm.1 |
NM_174993.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003869.mm.1 |
TC0X00000614.mm.1 |
MTA TC |