Assay Details
| Gene Symbol: Fmr1nb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166619.1 | NP_001160091.1 | 2 - 3 | 478 | 102 | |
| GenBank mRNA | AK076190.1 | - | 2 - 3 | 436 | 102 | |
| BC099411.1 | - | 2 - 3 | 478 | 30788714 | 102 | |
| BC138087.1 | - | 2 - 3 | 459 | 8861106 | 102 | |
| BC171923.1 | - | 2 - 3 | 459 | 9053691 | 102 | |
More Information
Set Membership: |
|

Fmr1nb
Genomic Map